SARS-CoV Recombinant Nucleocapsid Protein (His Tag) (from Baculovirus Insect Cells)
Catalog # 103663-804
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:SARS
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:NP_828858.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:Nucleoprotein|coronavirus Nucleocapsid|cov np|ncov NP|coronavirus NP|Nucleocapsid|NP|coronavirus Nucleoprotein|novel coronavirus Nucleoprotein
- Protein/Peptide Name:NP-CoV
- Purity:>80% as determined by SDS-PAGE.
- Molecular Weight:The recombinant SARS-CoV nucleoprotein comprises 433 amino acids and has a predicted molecular mass of 47.5 kDa. The apparent molecular mass of the protein is approximately 47.1 kDa in SDS-PAGE under reducing conditions.
- Sequence:Met1-Ala422
- Shipping Temperature:Ambient
- Cat. No.:103663-804
Specifications
About this item
A DNA sequence encoding the SARS-CoV (isolate:Tor2) nucleoprotein (NP_828858.1) (Met1-Ala422) was expressed with a C-terminal polyhistidine tag.