Order Entry
Puerto Rico
ContactUsLinkComponent
Influenza A H5N1 (A/Egypt/2321-NAMRU3/2007) Recombinant Neuraminidase/NA (Active) Protein (from HEK293 Cells)
Influenza A H5N1 (A/Egypt/2321-NAMRU3/2007) Recombinant Neuraminidase/NA (Active) Protein (from HEK293 Cells)
Catalog # 103632-784
Supplier:  Sino Biological
CAS Number:  
Influenza A H5N1 (A/Egypt/2321-NAMRU3/2007) Recombinant Neuraminidase/NA (Active) Protein (from HEK293 Cells)
Catalog # 103632-784
Supplier:  Sino Biological
Supplier Number:  40045-VNAHC-100
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    H5N1
  • Size:
    100 U
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    Measured by its ability to cleave a fluorogenic substrate, 2'-(4-Methylumbelliferyl)-α-D-N-acetylneuraminic acid. The specific activity is > 100 U.
    One unit is defined as the amount of enzyme required to cleave 1 nmole of 2'-(4-Methylumbelliferyl)-α-D-N-acetylneuraminic acid per minute at pH 7.5 at 37 ℃.
  • Gene ID:
    ACJ53852.1
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    Neuraminidase
  • Protein/Peptide Name:
    NA
  • Molecular Weight:
    The recombinant influenza A H5N1 Neuraminidase comprises 449 amino acids and has a predicted molecular mass of 49.1 kDa.
  • Sequence:
    Met1-Lys449
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    103632-784

Specifications

About this item

A DNA sequence encoding the Influenza A virus (A/Egypt/2321-NAMRU3/2007(H5N1)) neuraminidase (ACJ53852.1) (Met1-Lys449) was expressed, the cell lysates are collected, and bio-activity was tested.