Human Recombinant coronavirus HKU1 (isolate N1) (HCoV-HKU1) Spike/S1 Protein (S1 Subunit, His Tag) (from HEK293 Cells)
Catalog # 103663-506
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:HCoV
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:YP_173238.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:ncov RBD|ncov s2|ncov spike|coronavirus s2|S|s1|Spike RBD|ncov s1|coronavirus spike|coronavirus s1|cov spike|RBD
- Protein/Peptide Name:Spike
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:The recombinant human coronavirus HKU1 (isolate N1) (HCoV-HKU1) spike protein (S1 Subunit, His Tag) consists of 756 amino acids with a predicted molecular mass of 85.8 kDa. As a result of high glycosylation, the apparent molecular mass of the recombinant protein is approximately 130 - 140 kDa in SDS-PAGE under reducing conditions.
- Sequence:Met1-Arg760
- Shipping Temperature:Ambient
- Cat. No.:103663-506
Specifications
About this item
A DNA sequence encoding the human coronavirus HKU1 (isolate N1) (HCoV-HKU1) spike protein (S1 Subunit) (YP_173238.1) (Met 1-Arg 760) was expressed with a polyhistidine tag at the C-terminus.