Human Recombinant IGFLR1/TMEM149 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the human IGFLR1 (NP_078936.1) (Met1-Pro159) was expressed with the Fc region of human IgG1 at the C-terminus.
Specifications
- Conjugation:Unconjugated
- Protein Function:Immunotherapy
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:NP_078936.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:TMEM149|U2AF1L4|IGFLR1
- Protein/Peptide Name:IGFLR1
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant human IGFLR1/Fc comprises 375 amino acids and has a predicted molecular mass of 41.4 kDa. The apparent molecular mass of the protein is approximately 58.9 and 47.6 kDa in SDS-PAGE under reducing conditions due to glycosylation.
- Sequence:Met1-Pro159
- Shipping Temperature:Ambient
Specifications
Frequently Bought Together