Human Recombinant OX-40L/TNFSF4/CD252 Protein (ECD, Fc Tag) (from HEK293 Cells)
Catalog # 103616-276
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:50 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method
- Reconstitution Instructions:A hardcopy of COA with reconstitution instruction is sent along with the products, please refer to it for detailed information
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:gp34|CD134L|Ox40l|OX-40L|TXGP1|Txgp1l|Ath1|Ath-1
- Protein/Peptide Name:TNFSF4
- Purity:>95% as determined by SDS-PAGE
- Molecular Weight:The recombinant human TNFSF4 consists of 412 amino acids and predicts a molecular mass of 46.6 kDa.
- Sequence:Gln51-Leu183
- Formulation:Lyophilized from sterile PBS, pH 7.4
- Shipping Temperature:Ambient
- Cat. No.:103616-276
Specifications
About this item
A DNA sequence encoding the human TNFSF4 (Gln51-Leu183) was expressed with the Fc region of human IgG1 at the N-terminus.