Human Recombinant Interleukin-32/IL-32 Protein (isoform alpha, His Tag) (from HEK293 Cells)
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the human IL32 isoform alpha (IL32A) (NP_001012651.1) precursor (Met 1-Lys 131) was expressed with a polyhistidine tag at the C-terminus.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:NP_001012651.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:TAIFd|IL-32alpha|TAIFa|TAIFc|IL32|IL-32beta|IL-32gamma|NK4|TAIF|TAIFb|IL-32delta
- Protein/Peptide Name:IL32
- Purity:>95% as determined by SDS-PAGE
- Molecular Weight:The recombinant human IL32A comprises 137 amino acids with a predicted molecular mass of 15.7 kDa. As a result of glycosylation, it migrates as an approximately 20 kDa band in SDS-PAGE under reducing conditions.
- Sequence:Met1-Lys131
- Shipping Temperature:Ambient
Specifications
Frequently Bought Together