Human Recombinant CD20/MS4A1 Protein (TrxA Tag) (from E. coli)
Catalog # 103621-722
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein Function:Immunotherapy
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:Please contact us for more information.
- Gene ID:NP_068769.2
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:CVID5|MS4A1|Bp35|S7|LEU-16|MS4A2|CD20|B1
- Protein/Peptide Name:MS4A1
- Purity:>80% as determined by SDS-PAGE
- Molecular Weight:The recombinant human MS4A1 consists of 218 amino acids and predicts a molecular mass of 23.9 kDa. It migrates as an approximately 28.2 kDa band in SDS-PAGE under reducing conditions.
- Sequence:Ile141-Ser188
- Shipping Temperature:Ambient
- Cat. No.:103621-722
Specifications
About this item
A DNA sequence encoding the human MS4A1 (NP_068769.2) (Ile141-Ser188) was expressed with a TrxA tag at the N-terminus.