Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant Serum Albumin/HSA/HAS/ALB Protein (from Yeast)
 
undefined
Human Recombinant Serum Albumin/HSA/HAS/ALB Protein (from Yeast)

 

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Yeast
  • Species:
    Human
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    Please contact us for more information.
  • Gene ID:
    NP_000468.1
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    PRO1341|PRO0883|ANALBA|FDAH|PRO0903
  • Protein/Peptide Name:
    ALB
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    The recombinant mature human ALB consists of 585 amino acids and predicts a molecular mass of 66.4 kDa.
  • Sequence:
    Arg19-Leu609
  • Shipping Temperature:
    Ambient