Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant MST4 Protein (GST Tag) (from Baculovirus Insect cells)
Human Recombinant MST4 Protein (GST Tag) (from Baculovirus Insect cells)
Supplier:  Sino Biological
Certificates
undefined
Human Recombinant MST4 Protein (GST Tag) (from Baculovirus Insect cells)
Supplier:  Sino Biological

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Baculovirus (Insect cells)
  • Species:
    Human
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    The specific activity was determined to be 15 nmol/min/mg using MBP as substrate.
  • Gene ID:
    NP_057626.2
  • Reconstitution Instructions:
    A hardcopy of COA with reconstitution instruction is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    MASK|MST4
  • Protein/Peptide Name:
    STK26
  • Purity:
    >95% as determined by SDS-PAGE
  • Molecular Weight:
    The recombinant human MST41/GST chimera consists of 641 amino acids and predicts a molecular mass of 73 kDa. It migrates as an approximately 65 kDa band in SDS-PAGE under reducing conditions.
  • Sequence:
    Met1-Pro416
  • Shipping Temperature:
    Ambient

Specifications

Related Information