Order Entry
United States
ContactUsLinkComponent
 
Human Recombinant PSGL-1/CD162 Protein (His & Fc Tag) (from HEK293 Cells)
  103621-282
 :  
undefined
Human Recombinant PSGL-1/CD162 Protein (His & Fc Tag) (from HEK293 Cells)
  103621-282
 :  10490-H03H-100
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    100 µg
  • Environmentally Preferable:
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Gene ID:
    AAC50061.1
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    Psgl-1|Selp1|CD162|Psgl1|Selpl
  • Protein/Peptide Name:
    SELPLG
  • Purity:
    >95% as determined by SDS-PAGE
  • Molecular Weight:
    The recombinant human PSGL-1/Fc is a disulfide-linked homodimer after removal of the signal peptide and the prppeptide. The reduced monomer consists of 526 amino acids and has a predicted molecular mass of 57.1 kDa. In SDS-PAGE under reducing conditions, the apparent molecular mass of rh PSGL-1/Fc monomer is approximately 110-120 kDa due to glycosylation.
  • Sequence:
    Met1-Val295
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    103621-282

 

 

A DNA sequence encoding the extracellular domain (Met1-Val295) of human PSGL-1 precursor (AAC50061.1) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.