CBL0137 ≥95%
Supplier: Cayman Chemical Company
Certificates
About this item
CAS Number:1197996-80-7
:
Formula:C₂₁H₂₄N₂O₂
MDL Number:MFCD18071578
:336.4 g/mol
Storage Temperature:Freezer
Specification Test Results
- Formulation:A crystalline solid
- Purity:≥95%
- Stability:≥ 2 years
Specification Test Results
Frequently Bought Together