Order Entry
Puerto Rico
ContactUsLinkComponent
CELLvo™ Human Chondrocytes (HC-a) from Human articular cartilage, StemBioSys, Inc. (SBS)
CELLvo™ Human Chondrocytes (HC-a) from Human articular cartilage, StemBioSys, Inc. (SBS)
Supplier:  StemBioSys
Certificates
undefined
CELLvo™ Human Chondrocytes (HC-a) from Human articular cartilage, StemBioSys, Inc. (SBS)
Supplier:  StemBioSys

Specifications

  • Storage Conditions:
    CELLvo™ HCs should be stored in liquid nitrogen vapor and CELLvo™ XF-Matrix should be stored at 2-8° upon
  • Shipping Conditions:
    CELLvo™ HCs arrive on dry ice . HCs should be stored in liquid nitrogen upon arrival.
  • Storage Buffer:
    CryoStor®CS5
  • Tested Applications:
    in vitro vessel formation and cell phenotype

Specifications