Flow Control Adapters, PTFE Metering Stopcock, 90°, Chemglass
Supplier: Chemglass
Certificates
About this item
Used to regulate the flow of gases or liquids into reaction systems.
- Equipped with a 2 mm PTFE stopcock
- Internal needle valve permits fine control of gases or liquids
- For replacement stopcock, see CG-434-70
Stopcock has a bore of 2 mm, and has an internal needle valve which permits fine control of gases or liquids added to reactions. Hose connection has an O.D. of 8m m at the middle serration.