ContactUsLinkComponent
First Aid Only™ First Aid Tape, Acme United
Catalog # 76287-314
Supplier:  First Aid Only
undefined
First Aid Only™ First Aid Tape, Acme United
Catalog # 76287-314
Supplier:  First Aid Only
Supplier Number:  G634
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    First Aid Tape
  • Size:
    1/2" × 5 yards
  • Environmentally Preferable:
  • Cat. No.:
    76287-314
Specifications
About this item
First aid tape tears easily, yet is strong enough for securing catheters, tubing and dressings.
  • Tears easily for hassle-free taping and removal
  • Durable and comfortable
  • Molds to body contours
  • Remove without pain or shredding
The adhesive keeps the tape securely in place, but it is also easy to remove without pain and without shredding of the tape.