Aven 2-in-1 Soldering/Desoldering Station, Aven
76289-586
: Avantor
- Description:2-in-1 Soldering/De-Soldering Station
- Environmentally Preferable:
- Cat. No.:76289-586
200W Soldering Iron and a 130W De-Soldering gun and replaceable tips.
- Separate controls for soldering iron and desoldering gun
- Clear LCD displays
- Shows temperature for each
Includes a stand with compartments for the solder iron and de-soldering gun, a cleaning coil, and a recessed tray with a cleaning sponge.