Order Entry
United States
ContactUsLinkComponent
RtcB Ligase
RtcB Ligase
Catalog # 103307-256
Small Business Enterprise
CAS Number:  
undefined
RtcB Ligase
Catalog # 103307-256
Supplier Number:  M0458S
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Concentration:
    15 µM
  • Application:
  • Description:
    RtcB ligase
  • Environmentally Preferable:
  • Cat. No.:
    103307-256
  • No. of reactions:
    25
  • Supplier no.:
    M0458S

Specifications

About this item

RtcB Ligase is expressed as a His-tagged fusion in E. coli.

  • This is an Enzyme for Innovation (EFI)
  • These enzymes have interesting properties and unique specificities

RtcB Ligase from E. coli joins single stranded RNA with a 3'-phosphate or 2',3'-cyclic phosphate to another RNA with a 5' -hydroxyl. Ligation requires both GTP and MnCl₂ and proceeds through a 3'-guanylate intermediate. For substrates with a 2',3'-cyclic phosphate end, hydrolysis to a 3'-phosphate precedes 3' end activation with GMP and ligation.