Anti-Rabbit Detection Module, Protein Simple
Catalog # MSPP-DM001
Supplier: PROTEIN SIMPLE MS
CAS Number:
Specifications
- Description:Anti-Rabbit Detection Module
- Cat. No.:MSPP-DM001
- Supplier no.:DM-001
Specifications
About this item
Use with a Jess/Wes or Sally Sue/Peggy Sue Separation Module.
Includes Luminol-S, Peroxide, Antibody Diluent 2, Anti-Rabbit Secondary Antibody, Streptavidin-HRP.