About this item
CryoStor® CS10 is a uniquely formulated, serum-free, animal component-free and defined cryopreservation medium containing 10% dimethyl sulfoxide (DMSO). Designed to preserve cells in low temperature environments (-80 °C to -196 °C).
- Ready-to-use
- Serum free, protein free
- Animal component free
- cGMP manufactured with USP grade/highest quality components
- FDA master file
- Sterility, endotoxin, and cell-based quality control testing
CryoStor® CS10 provides a safe, protective environment for cells and tissues during the freezing, storage, and thawing processes. It is recommended for the cryopreservation of hepatocytes, tissue samples, human peripheral blood, human mesenchymal stem cells, human embryonic stem (ES) and induced pluripotent stem (iPS) cells, and other extremely sensitive cell types.