Hytrel® Lined PVC Tubing, Newage Industries
Supplier: NewAge Industries
Certificates
About this item
Hytrel® lined PVC tubing is designed to offer the flexibility and durability of PVC tubing with a liner of inert Hytrel, made by DuPont, for additional resistance to oils, resists U.V. exposure, ozone, high temperatures and gas permeability.
- Does not contain plasticizers
- Low permeability to refrigerant gases, hydrocarbons and moisture
- Superior barrier and dielectric characteristics
- Odorless, tasteless, and inert
- Ethylene oxide (EtO) sterilizable
- Hytrel® liner is REACH compliant
Polyester liner offers internal resistance to chemical attack, abrasion and contamination, along with a high degree of resistivity to the adherence of particulates, making exceptionally suited for use in air sampling applications. Made from FDA-compliant ingredients and suitable for food contact.
Certifications: Certified for CFR Title 21 Section 177.1550 and CFR Title 21 Section 177.2600.