Square Cell with Cap, Malvern Instruments
Square Cell with Cap (12 mm O.D.)
Catalog # MSPP-PCS8501
Marketsource
Supplier: MALVERN PANALYTICAL MS
Supplier #: PCS8501
Square Cell with Cap, Malvern Instruments
Catalog # MSPP-PCS8501
Supplier: MALVERN PANALYTICAL MS
Marketsource
Jump to:
Product Details & Documents
Square glass cell for size measurements.
- With circular aperture and stopper. Not suitable for use with the 'dip' electrophoresis electrodes.
Compatible with Zetasizer 1000/ 3000/ 3000HS, HPPS, Nano series and Zetasizer µV.
Specifications
- Description:Square Cell with Cap (12 mm O.D.)