CappWash™ Microplate Washer, Pipette.com
About this item
CappWash plate washer is an ideal ELISA plate washer for small-scale work. Simple, compact and inexpensive. Each tube consists of an inner tube that releases the distilled water washing solution into the wells for rinsing while the outer tube vacuums away the liquid upon completion of the washing procedure.
- Manual ELISA plate washer
- Simple & robust design
- Easy to operate
- Fully autoclavable
- Highly resistant to chemicals
- equires no programming nor electrical wiring connections
3 Options Available Below
Product Details & Documents
CappWash plate washer is an ideal ELISA plate washer for small-scale work. Simple, compact and inexpensive. Each tube consists of an inner tube that releases the distilled water washing solution into the wells for rinsing while the outer tube vacuums away the liquid upon completion of the washing procedure.
- Manual ELISA plate washer
- Simple & robust design
- Easy to operate
- Fully autoclavable
- Highly resistant to chemicals
- equires no programming nor electrical wiring connections
Product Family Options
Product Information