Order Entry
Puerto Rico
ContactUsLinkComponent
Sodium Phosphate Buffer, MP Biomedicals
Sodium Phosphate Buffer, MP Biomedicals
Catalog # IC116560605
Supplier:  MP Biomedicals
CAS Number:  
Sodium Phosphate Buffer, MP Biomedicals
Catalog # IC116560605
Supplier:  MP Biomedicals
Supplier Number:  116560605
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Storage Conditions:
    Store at Room Temperature(15 - 30 °C).
  • Cat. No.:
    IC116560605
  • pH:
    7.7 - 8.3

Specifications

About this item

Sodium phosphate is a reagent with very high buffering capacity that is widely used in molecular biology, biochemistry, and chromatography.

  • It occurs in several forms: monobasic (NaH₂PO₄), dibasic (NaH₂PO₄), and tribasic (Na₃PO₄)
  • Sodium phosphate buffer is a component of FastDNA® SPIN Kits
  • Instrument compatibility with FastPrep-24; FastPrep 120; FastPrep-96; SuperFastPrep-1
  • Component of FastDNA® SPIN Kit

Application includes: Sample preparation; sample lysis; nucleic acid isolation and purification; molecular biology; gene expression; microbiological analysis; gene function; gene regulation.

Ordering information: FastPrep® Instrument or alternative high speed disruptor, appropriate lysing matrix tubes, microcentrifuge, pipettmen, nucleic acid purification kit and/or chloroform, 100% ethanol, 75% ethanol, 1.5 or 2.0 ml nuclease-free microcentrifuge tubes, sgarose, gel loading dye and RNA size marker.