Order Entry
Puerto Rico
ContactUsLinkComponent
GeneClean® Turbo Kit, MP Biomedicals
GeneClean® Turbo Kit, MP Biomedicals
Catalog # 76019-990
Supplier:  MP Biomedicals
CAS Number:  
undefined
GeneClean® Turbo Kit, MP Biomedicals
Catalog # 76019-990
Supplier:  MP Biomedicals
Supplier Number:  111102400
CAS Number:  

Specifications

  • Sample type:
    Gel matrix
    Liquids
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Description:
    GENECLEAN® Turbo Kit
  • Size:
    100 preps
  • Sample Size:
    Up to ~100 mg gel slice or up to ~100 µl DNA solution
  • Cat. No.:
    76019-990
  • Includes:
    GeneClean® Turbo Salt Solution(55 ml), GeneClean® Turbo Wash Concentrate(5 ml), GeneClean® Turbo Cartridges(50 cartridges), GeneClean® Turbo Catch Tubes(50 tubes), GeneClean® Turbo Elution Solution(3.3 ml), GeneClean® Turbo GNomic Salt Solution(14 ml)

Specifications

About this item

GeneClean® Turbo kit is used for purification of DNA fragments of sizes 0.1 to 300 kb from agarose gels, PCR reactions and other enzymatic solutions. Purified DNA can be used in enzymatic reactions, conventional or automated sequencing, cloning and microarray analysis.

  • Isolate DNA from TAE or TBE buffered agarose gels, PCR reactions, and other enzymatic solutions
  • Save time with patented Turbo fi lter design
  • Process with either vacuum or centrifugation
  • Purify larger fragments due to less manipulation of bound DNA
  • No carryover of matrix into DNA solution
  • Fastest, most flexible kit

GeneClean® technology simplifies the process of purifying DNA into three easy steps: bind, wash, and elute. Ethanol precipitation is never required, and purified DNA is immediately ready for a wide variety of downstream applications.

GeneClean® Turbo Kit is a flexible kit designed for purification of DNA fragments of sizes 0.1 kb to 300 kb from TAE or TBE buffered agarose gels, PCR reactions and other enzymatic solutions. It purifies larger fragments due to less manipulation of bound DNA. Process with either vacuum or centrifugation and save time with patented turbo filter design.

Isolate DNA from: TAE or TBE buffered agarose gels, enzymatic reactions, buffered solutions (desalting), PCR products from templates, labeling reactions.

Application includes: Nucleic Acid Isolation and Purification.

All GeneClean® Turbo Kits are shipped ambiently. Store all reagents at room temperature (15 - 30 °C).