CENTA™ β-Lactamase Substrate ≥98% (by HPLC), Calbiochem®
80050-988
:
- Pack type:Plastic ampoule
- Size:25 mg
- Cat. No.:80050-988
- Supplier no.:219475-25MG
- Purity (HPLC):≥98%
:
:535.57 g/mol
:Freezer
:80072-86-2
:MFCD00131819
:C₂₁H₁₇N₃O₈S₃