Triethylammonium Bicarbonate Buffer, Volatile Buffer for HPLC, Fluka™, Honeywell Research Chemicals
Supplier: Honeywell Research Chemicals
Certificates
About this item
Triethylammonium bicarbonate buffers are volatile buffer for HPLC.
- Non-volatile matter: Max. 0.05 %
- pH (20 °C): 8.4 - 8.6
- Absorbance at 260 nm: Max. 0.04
- Absorbance at 280 nm: Max. 0.04