Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Safe-Lock Tubes, Eppendorf

1.5 ml, PP, Assorted, 30000×g
Catalog # MSPP-022363328
Marketsource
 :  022363328
Safe-Lock Tubes, Eppendorf
Catalog # MSPP-022363328
Marketsource
Safe-Lock Tubes, Eppendorf

1.5 ml, PP, Assorted, 30000×g

Price
$90.20
Log in for availability
Pack of 500
 
Product Details & Documents

These safe-lock tubes protect the sample with excellent centrifugation stability, feature optimized sealing properties, and reliably prevent sample evaporation.

  • Hinged lid prevents unintentional lid opening during incubation and storage
  • Precise lid sealing for minimal evaporation rates during long-term storage
  • Large frosted lid and surface on the side for easy labeling
  • Guaranteed functionality from ‒86 to 100 °C
  • Autoclavable when open (121 °C, 20 min)

Provides high resistance to chemicals, mechanical stress and temperature extremes. Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results. g-Safe® with exceptional centrifugation stability up to 30000×g and prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples.

Specifications
  • Capacity:
    1.5 ml
  • Color:
    Assorted
  • Material:
    PP
  • Max. rcf:
    30000×g
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.