Order Entry
Puerto Rico
ContactUsLinkComponent
Multi-Ion Standard 100 ppm each Ca²+, Mg²+, Na+, NH4+, Li+, K+ for Ion Chromatography, Ricca Chemical Company
Multi-Ion Standard 100 ppm each Ca²+, Mg²+, Na+, NH4+, Li+, K+ for Ion Chromatography, Ricca Chemical Company
Catalog # RCRMSTD200120A
Supplier:  Ricca Chemical
CAS Number:  
undefined
Multi-Ion Standard 100 ppm each Ca²+, Mg²+, Na+, NH4+, Li+, K+ for Ion Chromatography, Ricca Chemical Company
Catalog # RCRMSTD200120A
Supplier:  Ricca Chemical
Supplier Number:  RMSTD200-120A
CAS Number:  

Specifications

  • Concentration:
    100 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Poly Bottle
  • Size:
    120 ml
  • Cat. No.:
    RCRMSTD200120A
  • Accuracy:
    0.5 mg/L
  • Contents:
    NH₄+, Ca2+, K+, Li+, Mg2+, Na+,

Specifications

About this item