Tech-Med® Digital Timer, Dukal Corporation®
Catalog # 76235-396
Supplier: Dukal
Specifications
- Description:Digital timer
- Cat. No.:76235-396
Specifications
About this item
Pocket size, with a spring loaded clip and magnet.
- Counts down and up from the max. time of 99 minutes
- 0.5" number display
Delivery: Includes battery.