Order Entry
Puerto Rico
ContactUsLinkComponent
Nippon Genetics Bambanker HRM Cell Freezing Medium for iPS and ES Cells
Nippon Genetics Bambanker HRM Cell Freezing Medium for iPS and ES Cells
Catalog # 102407-918
Supplier:  Bulldog Bio
CAS Number:  
undefined
Nippon Genetics Bambanker HRM Cell Freezing Medium for iPS and ES Cells
Catalog # 102407-918
Supplier:  Bulldog Bio
Supplier Number:  BBH01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    iPS and ES cell freezing media
  • Size:
    20 ml
  • Cat. No.:
    102407-918
  • Supplier no.:
    BBH01

Specifications

About this item

Long term storage of ES and iPS cells at −80 °C or in liquid nitrogen.

  • Up to 3× better recovery rates
  • No step-wise freezing