Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Hybridoma Cell Freezing Media
- Size:20 ml
- Cat. No.:102407-916
- Supplier no.:BBD01
Specifications
About this item
Long-Term Storage of Hybridomas at -80 °C or in Liquid Nitrogen.
- Up to 3× Better Recovery Rates
- No Step-wise Freezing
There are certain cell types, such as hybridomas, which exhibit increased sensitivity to “over” manipulations. These cells often exhibit higher death rates or unwanted differentiation after long-term term storage and freeze thaw cycling.
It is for these types of cells that Bambanker DIRECT was created – to provide tight environmental control. By eliminating the centrifugation step, cells can now be frozen for long-term storage immediately upon addition of the novel cryoprotectant.
Just add Bambanker DIRECT, one-to-one, with your cells in media and directly place in freezer. And like with the original Bambanker, the DIRECT version has no serum, so it is great for cells which animal-derived serum may be an issue.