Specifications
- Description:Pre-measured Agarose Tablets
- Size:200 tablets
- Cat. No.:102407-908
- Supplier no.:AG05
Specifications
About this item
No Weighing Required.
Just add one tablet to 50 ml of gel running buffer and heat—the result is a 1% agarose gel. It’s that simple. With our FastGene Agarose Tablets there’s no powder to weigh and no mess to make!