Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant IRF3 (from E. coli)
Human Recombinant IRF3 (from E. coli)
Catalog # 76292-902
CAS Number:  
undefined
Human Recombinant IRF3 (from E. coli)
Catalog # 76292-902
Supplier Number:  22811-100
CAS Number:  

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Storage Conditions:
    –80 °C
  • Shipping Conditions:
    –80 °C, dry ice
  • Enzyme Name:
    IRF3
  • Purity:
    ≥85% as estimated by SDS-PAGE
  • Molecular Weight:
    49.6 kDa
  • Formulation:
    50 mM HEPES, pH 8.0, 150 mM sodium chloride, and 10% glycerol
  • Cat. No.:
    76292-902

Specifications

About this item

Interferon regulatory factor 3 (IRF3) is a member of the IRF family. It plays a crucial role in activation of innate immunity and inflammation in response to viral infection, functioning as a molecular switch for antiviral activity. Double-stranded RNA generated during a viral infection leads to IRF3 activation by serine/threonine phosphorylation by TBK1 or IKBKE kinases, which induces a conformational change leading to its dimerization, nuclear localization, and association with CREBBP. The complex, known as DRAF1, activates transcription interferons alpha and beta, as well as other interferon-induced genes. These genes play a critical role in type 1 interferon (IFN)-dependent immune response. A recent study shows phosphorylation of innate immune adaptor proteins MAVS, STING, and TRIF recruits and specifies IRF3 phosphorylation and activation by the serine/threonine-protein kinase TBK1, thereby inducing the production of type-1 interferons.

Used for: Apoptosis, Transcription Factors, Signal Transduction, Innate Immunity