Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant CdnP (from M. Tuberculosis strain ATCC 25618/H37Rv)
Human Recombinant CdnP (from M. Tuberculosis strain ATCC 25618/H37Rv)
Catalog # 76292-930
CAS Number:  
undefined
Human Recombinant CdnP (from M. Tuberculosis strain ATCC 25618/H37Rv)
Catalog # 76292-930
Supplier Number:  22809-100
CAS Number:  

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    M. tuberculosis strain ATCC 25618/H37Rv
  • Species:
    Human
  • Size:
    100 µg
  • Storage Conditions:
    –80 °C
  • Shipping Conditions:
    –80 °C, dry ice
  • Enzyme Name:
    CdnP
  • Purity:
    ≥70% estimated by SDS-PAGE
  • Molecular Weight:
    52.71 kDa
  • Formulation:
    50 mM HEPES, pH 8, containing 150 mM sodium chloride
  • Cat. No.:
    76292-930

Specifications

About this item

Cyclic di-nucleotide phosphodiesterase (CdnP, also known as Rv2837c) is a soluble, stand-alone phosphodiesterase in regulating cyclic dinucleotide signaling during intracellular infections of M. tuberculosis{36095}. Studies have found that CdnP is capable of hydrolyzing cyclic di-adenosine monophosphate (c-di-AMP; Item No. 17753) into two AMPs as a strategy to avoid activation of the innate immune response of the host. M. tuberculosis infection leads to cytosolic release of c-di-AMP, which is recognized by stimulator of interferon genes (STING) and subsequently triggers type I interferon response. Other studies have shown that deletion or inhibition of CdnP attenuates the M. tuberculosis virulence both in vitro and in vivo.

Used for: Antivirals, Autoimmunity, Infectious Disease, Innate Immunity, Tuberculosis Research