Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-KS Mouse Monoclonal Antibody [clone: R-10G]
Anti-KS Mouse Monoclonal Antibody [clone: R-10G]
Catalog # TCA2968-VIAL
Supplier:  TCI America
Anti-KS Mouse Monoclonal Antibody [clone: R-10G]
Catalog # TCA2968-VIAL
Supplier:  TCI America
Supplier Number:  A2968-1VIAL

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Keratin Sulfate
  • Antigen Symbol:
    KS
  • Clonality:
    Monoclonal
  • Clone:
    R-10G
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Western Blot:
    Yes
  • Size:
    0.1 mg
  • Format:
    Ascites Fluid
  • Storage Buffer:
    Phosphate buffered-saline (PBS)
  • Storage Temperature:
    Store at -20 ºC to -80 ºC (Preferably at -80 ºC). It is not recommended storing at 4 ºC. Avoid repeated freezing and thawing.
  • Shipping Temperature:
    -80°C
  • Immunogen:
    Purified mouse monoclonal antibody/IgG1
  • Cat. No.:
    TCA2968-VIAL
  • Packaging:
    Vial

Specifications

About this item

The R-10G monoclonal antibody specifically binds to human induced pluripotent stem cells(hiPS) and human embryonic stem cells(hES), is useful tool for the evaluation of hiPS/hES cells. The epitope of R-10G is a unique form of the keratan sulfate associated with podocalyxin on hiPS/ES cells, which lacks oversulfated structures. The epitope is also expressed in the adult human and mouse brains.

Type: Primary
Antigen: KS
Clonality: Monoclonal
Clone: R-10G
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: