Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant cGAS (from E. coli)
Human Recombinant cGAS (from E. coli)
Catalog # 76292-894
CAS Number:  
undefined
Human Recombinant cGAS (from E. coli)
Catalog # 76292-894
Supplier Number:  25001-100
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Storage Conditions:
    –80 °C
  • Shipping Conditions:
    –80 °C, dry ice
  • Enzyme Name:
    cGAS
  • Enzyme Synonyms:
    C6orf150, 2'3'-cGAMP Synthease, cGAMP Synthase, h-cGAS, Cyclic GMP-AMP Synthase, Mab-21 domain containing protein 1, MB21D1
  • Purity:
    ≥90% estimated by SDS-PAGE
  • Molecular Weight:
    44.6 kDa
  • Formulation:
    50 mM HEPES, pH 8.0, 150 mM sodium chloride, 1 mM DTT, and 10% glycerol
  • Cat. No.:
    76292-894

Specifications

About this item

Cyclic GMP-AMP (cGAMP) synthase (cGAS) (161-522) is a truncated form of cGAS (Item No. 22810) that contains the nucleotidyltransferase and Mab21 domains. cGAS is a nucleotidyltransferase located in the cytosol that acts as a cytosolic DNA sensor to detect foreign DNA from microbial pathogens as part of the innate immune response. Upon binding to cytosolic DNA, cGAS produces the cyclic dinucleotide second messenger cGAMP, which activates stimulator of interferon genes (STING), leading to activation of the type I interferon (IFN) pathway. In vitro, fibroblasts, macrophages, and dendritic cells isolated from cGAS knockout (cGAS-/-) mice do not produce type I IFNs following DNA transfection or DNA virus infection. Similarly, cells containing a frame-shift mutation in the cGAS locus fail to mount an immune response to HIV and other retroviruses. In vivo, cGAS-/- mice infected with herpes simplex virus 1 (HSV-1) have lower levels of IFN-α and IFN-β, shorter survival times, and higher post-mortem levels of HSV-1 in the brain.

  • UniProt: Q8N884

Used for: Innate Immunity, Infectious Disease, Intracellular Signaling