Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant cGAS (from E. coli)
Catalog # 76292-892
CAS Number:  
undefined
Human Recombinant cGAS (from E. coli)
Catalog # 76292-892
Supplier Number:  22810-100
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Storage Conditions:
    –80 °C
  • Shipping Conditions:
    –80 °C, dry ice
  • Enzyme Name:
    cGAS
  • Enzyme Synonyms:
    C6orf150, 2'3'-cGAMP Synthease, cGAMP Synthase, h-cGAS, Cyclic GMP-AMP Synthase, EC: 2.7.7.86, Mab-21 domain containing protein 1, MB21D1
  • Purity:
    ≥90% estimated by SDS-PAGE
  • Molecular Weight:
    61 kDa
  • Formulation:
    50 mM HEPES, pH 8.0, 300 mM sodium chloride, and 10% glycerol
  • Cat. No.:
    76292-892

Specifications

About this item

Cyclic GMP-AMP (cGAMP) synthase (cGAS) is a nucleotidyltransferase located in the cytosol that acts as a cytosolic DNA sensor to detect foreign DNA from microbial pathogens as part of the innate immune response. Upon binding to cytosolic DNA, cGAS produces the cyclic dinucleotide second messenger cGAMP, which activates (STING), leading to activation of the type I interferon (IFN) pathway. In vitro, fibroblasts, macrophages, and dendritic cells isolated from cGAS knockout (cGAS-/-) mice do not produce type I IFNs following DNA transfection or DNA virus infection. Similarly, cells containing a frame-shift mutation in the cGAS locus fail to mount an immune response to HIV and other retroviruses. In vivo, cGAS-/- mice infected with herpes simplex virus 1 (HSV-1) have lower levels of IFN-α and IFN-β, shorter survival times, and higher post-mortem levels of HSV-1 in the brain.

  • UniProt: Q8N884

Used for: Transcription Factors, Innate Immunity, Infectious Disease, Autoimmunity, Antivirals