Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Ethidium monoazide bromide (EMA) ≥95% (by HPLC)

5 mg
Catalog # 103011-138
Marketsource
Supplier:   Anaspec
 :  AS-83214
Ethidium monoazide bromide (EMA) ≥95% (by HPLC)
Catalog # 103011-138
Supplier:  Anaspec
Marketsource
CAS Number:  58880-05-0
Ethidium monoazide bromide (EMA) ≥95% (by HPLC)

5 mg

CAS Number:  58880-05-0
Price
$324.46
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

A fluorescent photoaffinity label that couples covalently to nucleic acids upon light irradiation

  • Spectral properties: Abs/Em = 462/625 nm
  • Solvent system: Water
  • Storage Temperature: -20°C desiccated and protected from light
  • Shipping condition: Blue ice
  • CAS Number:58880-05-0
  • Formula:C₂₁H₁₈BrN₅
  • Molecular Weight:420.305 g/mol
  • Storage Temperature:Freezer
Specifications

Specifications

  • Size:
    5 mg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.