Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
6-Carboxy-2',4,4',5',7,7'-hexachlorofluorescein succinimidyl ester (6-HEX SE)

6-Carboxy-2',4,4',5',7,7'-hexachlorofluorescein succinimidyl ester (6-HEX SE)

5 mg
Catalog # 103010-794
Marketsource
Supplier:   Anaspec
Supplier #:  AS-81020
6-Carboxy-2',4,4',5',7,7'-hexachlorofluorescein succinimidyl ester (6-HEX SE)
Catalog # 103010-794
Supplier:  Anaspec
Marketsource
CAS Number:  2129651-79-0
6-Carboxy-2',4,4',5',7,7'-hexachlorofluorescein succinimidyl ester (6-HEX SE)

5 mg

CAS Number:  2129651-79-0
Price
$420.92
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

6-HEX, SE is a popular amino-reactive fluorescent probe that is widely used in nucleic acid sequencing and related research.

  • Spectral properties: Abs/Em = 533/550 nm
  • Solvent system: DMF or DMSO
  • Storage Temperature: -20°C desiccated and protected from light
  • Shipping condition: Blue ice
Synonyms:  6-HEX, NHS ester
  • CAS Number:2129651-79-0
  • Formula:C₂₅H₉Cl₆NO₉
  • Molecular Weight:680.06 g/mol
  • Storage Temperature:Freezer
Specifications

Specifications

  • Size:
    5 mg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.