Order Entry
Puerto Rico
ContactUsLinkComponent
 
ARP
Catalog # 103003-296
Supplier:  Anaspec Inc
CAS Number:  
ARP
Catalog # 103003-296
Supplier:  Anaspec Inc
Supplier Number:  AS-60645
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    10 mg
  • Cat. No.:
    103003-296
  • Supplier no.:
    AS-60645

Specifications

About this item

 :
 :445.42 g/mol
Storage Temperature:Freezer
Formula:C₁₄H₂₂F₃N₅O₆S
MDL Number:MFCD03452814