Order Entry
Puerto Rico
ContactUsLinkComponent
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I)
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I)
Catalog # 102998-068
Supplier:  Anaspec Inc
CAS Number:  
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I)
Catalog # 102998-068
Supplier:  Anaspec Inc
Supplier Number:  AS-20151
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    1 g
  • Cat. No.:
    102998-068
  • Supplier no.:
    AS-20151

Specifications

About this item

 :
 :389.381 g/mol
Storage Temperature:Freezer
CAS Number:3326-32-7
Formula:C₂₁H₁₁NO₅S
MDL Number:MFCD00005063