ContactUsLinkComponent
Chromogenic Western Blot Kit (Anti-Mouse), Enzo Life Sciences
Chromogenic Western Blot Kit (Anti-Mouse), Enzo Life Sciences
Catalog # 76288-290
Supplier:  Enzo Life Sciences
CAS Number:  
Chromogenic Western Blot Kit (Anti-Mouse), Enzo Life Sciences
Catalog # 76288-290
Supplier:  Enzo Life Sciences
Supplier Number:  KIT182-0001
CAS Number:  
Specifications
  • Description:
    Chromogenic Western Blot Kit (Anti-Mouse)
  • Size:
    1 Kit
  • Cat. No.:
    76288-290
Specifications
About this item
Chromogenic Western Blot kit with increased signal and sensitivity.
  • More sensitive detection - able to detect lower concentrations of protein
  • Easy to use - all reagents are ready-to-use or ready-to-dilute, saving precious time
  • Easily adaptable - works with a wide range of mouse-derived proteins
The Chromogenic Western Blot Kit (anti-mouse) can be used to identify primary antibodies on a Western Blot using immunodetection. The kit contains a complete reagent set, with optimized ready-to-use or ready-to-dilute reagents.