Order Entry
United States
ContactUsLinkComponent
Magid® EconoWear® Polyethylene Bib Apron, Magid Glove
Magid® EconoWear® Polyethylene Bib Apron, Magid Glove
Catalog # 69100-091
Supplier:  Magid Glove
Magid® EconoWear® Polyethylene Bib Apron, Magid Glove
Catalog # 69100-091
Supplier:  Magid Glove
Supplier Number:  117W

Specifications

  • Sterile:
    +
  • Garment application:
    Industrial
  • Gender:
  • Color:
    White
  • Environmentally Preferable:
  • Fabric:
    PE
  • Cat. No.:
    69100-091
  • W×L:
    28×45"
  • Supplier no.:
    117W

Specifications

About this item

Magid® EconoWear® White Disposable PE Apron with binding ties and accepted for food handling.

Certifications: Meets USDA.