Palladium 20% on activated carbon powder (Pearlman Catalyst) wetted with water 24 µm for use as a catalyst, unreduced , BASF Catalyst
Supplier: Spectrum Chemical Mfg. Corp.
Certificates
About this item
CAS Number:7440-05-3
:
Formula:Pd
MDL Number:MFCD00011167
:106.420 g/mol
Storage Temperature:Ambient