Evergreen Scientific Slide Moisture Chambers for IHC Staining, Newcomer Supply
Supplier: Newcomer Supply
Certificates
About this item
The slide moisture chamber is ideal for IHC staining and keeping the slides well separated, maintaining moisture and watching the reaction.
- Divided into 10 individual compartments
- Slides are placed on 4 pedestal posts to raise the surface above water level
- Air tight design
- Available in clear and black
- Overall dimensions 81/4×7×11/4 "
- Temperature rating from –10 to 70 °C
The slide moisture chamber is stackable and made of heavy duty plastic construction. The clear slide staining tray chamber allows easy visualization of the reaction and the black slide staining tray is effective for light sensitive chemistry.
There are 8 barrier dividers in the empty spaces between compartments for complete isolation when lid is closed.