Airfree® Schlenk Double Bubbler, Chemglass
80064-568
Small Business Enterprise
- Description:Airfree® Schlenk Double Bubbler
- Environmentally Preferable:
- Cat. No.:80064-568
- Supplier no.:AF-0513-D-21
Double bubbler with two oil reservoirs.
- Used for monitoring gas flow to reactions while providing a vent to the atmosphere
- Reservoir head prevents oil from being sucked back into the system
Bubbler is approximately 125 W×135 mm H. Hose connections are 9 mm O.D. at the middle serration.