Order Entry
Puerto Rico
ContactUsLinkComponent
Spin Concentrators, 50K MWCo, Agilent Technologies
Spin Concentrators, 50K MWCo, Agilent Technologies
Catalog # 76204-860
undefined
Spin Concentrators, 50K MWCo, Agilent Technologies
Catalog # 76204-860
Supplier Number:  5188-5202

Specifications

  • Description:
    Spin Concentrators, 50K MWCo
  • Size:
    4 ml
  • Cat. No.:
    76204-860

Specifications

About this item

Spin concentrators are devices used to concentrate protein samples via low-speed centrifugation through MWCo filters into a collection tube.

  • Contains a polyethersulfone (PES) membrane that is compatible with biological fluids and most aqueous solutions with pore sizes of 50 kDa
  • Active membrane area of 2.0 cm2 or 4.0 cm2
  • Durable and designed to minimize membrane fouling and allow fast concentration, even with particles in the solution

Designed to fit a standard 15 ml conical collection tube or a 50 ml collection tube (not provided with concentrator). Designed to be used in either swing-bucket or fixed angle rotor centrifuge equipment at maximum 5000 g or 2000 g.