Airfree® Schlenk Trap Connection Adapter, Chemglass
Catalog # 80064-410
Supplier: Chemglass
Small Business Enterprise
Specifications
- Description:Airfree® Schlenk Trap Connection Adapter
- Cat. No.:80064-410
- Supplier no.:AF-0300-14
Specifications
About this item
Adapter used to connect the 80064-408 manifold to the 80064-400 trap assembly.
Adapter has a 0 to 20 mm Chem-Cap® valve with two #20 O-Ring joints.
Delivery: Supplied complete with two #214 Viton O-Rings.