Airfree® Schlenk Connecting Adapters, with Frit, Chemglass
Supplier: Chemglass
Certificates
About this item
Adapter has a coarse frit sealed between the #15 O-Ring joint and the standard taper inner joint.
- Coarse frit sealed between the joints
Frit permits filtration of large particles before introduction into the flask and also serves as a bump trap for reactions being performed in flasks while connected to the manifold.
Delivery: Supplied complete with one #116 Viton O-Ring.