Order Entry
United States
ContactUsLinkComponent
Airfree® Schlenk Double Bank Manifolds, Chemglass
Airfree® Schlenk Double Bank Manifolds, Chemglass
Catalog # 80064-408
Supplier:  Chemglass
Small Business Enterprise
undefined
Airfree® Schlenk Double Bank Manifolds, Chemglass
Catalog # 80064-408
Supplier:  Chemglass
Supplier Number:  AF-0300-13
Small Business Enterprise

Specifications

  • Description:
    Airfree® Schlenk 3-Port Vacuum Manifold, Double Bank
  • Environmentally Preferable:
  • Cat. No.:
    80064-408
  • Overall length:
    410 mm
  • Supplier no.:
    AF-0300-13

Specifications

About this item

Double bank, three-port manifold with 0 to 8 mm Chem-Vac™, Chem-Cap® valves for use on the (80060-052) portable cart.

  • Ports have 15 mm O-Ring joints and are spaced 100 mm center to center
  • Manifold has an overall length of 410 mm
  • Left side of maniflold has 20 mm O-Ring joints for the connection to (80064-410) adapters
  • Right side of manifold has 15 mm O-Ring joints for use with the gauge adapter and (80064-394) stopper

Valves face forward, permitting convenient operation of both vacuum and inert gas lines. Requires the use of an (80064-410) adapter for connection to the (80064-400) trap assembly at the left top bank of the manifold.

Please note: Manifold is not supplied with O-Rings.