Airfree® Probe Holders for Bioreactors, Chemglass
Catalog # 10034-268
Supplier: Chemglass
Small Business Enterprise
Specifications
- Description:Replacement Viton Compression Ring for 19 mm Probe
- Environmentally Preferable:
- Cat. No.:10034-268
- Supplier no.:1381-009
Specifications
About this item
Probe holder for 19 mm probes.
- Adjustable height with a compression fitting
Serve as replacement parts for Applikon® Biotechnology and Broadley-James® brands of bioreactors.